Learn More
CPC Scientific H-Arg-Lys-Ala-Ala-Pro-Ala-Thr-Gly-Gly-Val-Lys(Me3)-Lys-Pro-His-Arg-Tyr-Arg-Pro-Gly-Thr-Val-Lys(Biotin)-OH (trifluoroacetate salt) 0.25MG
SDP
Supplier:  CPC Scientific HIST064A
Encompass
This peptide is the fragment of Histone H3 (26-46) with a tri-methylated Lys36. The C-terminal contains a biotinylated Lys. It is associated with RNA polymerase II (Pol II) elongation in Saccharomyces cerevisiae.ONE-LETTER SEQUENCE: RKAAPATGGV-K(Me3)-KPHRYRPGTV-K(BIOTIN)MOLECULAR FORMULA: MOLECULAR WEIGHT:2644.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Baetz, K. et al. Mol Cell Biol 26, 7871 (2006)
Catalog No. 50-210-9514