Learn More
Abcam LYS6K2, S6 kinase inhibitor, 25MG
Supplier:  Abcam AB14619925MG
Encompass_Preferred
MW 445.42 g/mol. Selective, cell-permeable S6 kinase inhibitor. Blocks phosphorylation of p70 ribosomal S6 kinase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4850