Learn More
CPC Scientific H-Glu-Ile-Ala-Gln-Asp-Phe-Lys(Me3)-Thr-Asp-Leu-Arg-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HIST019B
Encompass
This peptide is the fragment of Histone H3 (73-83) tri-methylated at Lysine79. Histone lysine methylation is associated with transcriptional regulation. H3K79 can be methylated by Dot1 methyltransferase.ONE-LETTER SEQUENCE: EIAQDF-K(Me3)-TDLRMOLECULAR FORMULA: MOLECULAR WEIGHT:1378.5STORAGE CONDITIONS: -20 5CRESEARCH AREA: MiscellaneousREFERENCES: Bheda, P. et al. PNAS 109, E916 (2012)
Catalog No. 50-210-9452