Learn More
Cayman Chemical Lyso-Monosialoganglioside GM1
SDP
Supplier:  Cayman Chemical 24837500 UG
Encompass_Preferred
A ganglioside lacking the fatty acyl group; inhibits laminin-1-induced clustering of ganglioside GM1 in DRG cells (2.5 µM); does not protect against glutamate-induced neurotoxicity in primary neuronal cultures; elevated levels have been found in the brain and spinal cord of patients with infantile- and late-infantile-type GM1-gangliosidosis
Catalog No. 50-301-5793