Learn More
Medchemexpress LLC Lyso-PAF C-16 | 52691-62-0 | 99.8% | C24H52NO6P | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1415701MG
Encompass_Preferred
Lyso-PAF C-16 is a substrate for lysoplasmalogen (LysoPls)-specific phospholipase D (LysoPLD). It selectively acetylates with arachidonic acid and is intended for research use only. It has demonstrated cytotoxicity against various human cancer cell lines, including A549, HCT15, SK-MEL-3, SKOV3, and XF498.
  • Substrate for lysoplasmalogen (LysoPls)-specific phospholipase D (LysoPLD)
  • Selectively acetylates with arachidonic acid
  • Intended for research use only
  • Exhibits cytotoxicity against human A549 cells (ED50 5.5 μg/mL)
  • Exhibits cytotoxicity against human HCT15 cells (ED50 5.4 μg/mL)
  • Exhibits cytotoxicity against human SK-MEL-3 cells (ED50 5.4 μg/mL)
  • Exhibits cytotoxicity against human SKOV3 cells (ED50 4.9 μg/mL)
  • Exhibits cytotoxicity against human XF498 cells (ED50 4.9 μg/mL)
Catalog No. 50-003-56807