Learn More
Cayman Chemical Lyso-PAF C-16-d4 1mg
SDP
Supplier:  Cayman Chemical 3609061 MG
Encompass_Preferred
An internal standard for the quantification of PAF C-16 by GC- or LC-MS.
Catalog No. 50-291-3201