Learn More
Medchemexpress LLC Lyso-PAF C-18 | 74430-89-0 | MFCD00065889 | 98.0% | 509.70 g·mol⁻¹ | C26H56NO6P | 5 MG

Supplier: Medchemexpress LLC HY1415815MG
Lyso-PAF C-18 is a research intermediate used for the synthesis of C18-PAF and related platelet-activating factor derivatives. It is supplied as a purified lipid for organic synthesis and biological research and is intended for research use only.
- High purity (98.0%) for consistent experimental results.
- Molecular formula C26H56NO6P; molecular weight 509.70 g·mol⁻¹.
- Available in small laboratory quantities suitable for synthesis and screening.
- Powder stable at -20°C for long-term storage; in solvent stability limited over shorter periods.
- Certificate of analysis and safety data sheet available to support use.
- Useful intermediate for preparation of platelet-activating factor analogs.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.