Learn More
Medchemexpress LLC LysoPE(18:2/0:0) | 85046-18-0 | 99.6% | 477.57 g/mol | C23H44NO7P | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15491810MG
Encompass_Preferred
LysoPE(18:2/0:0) is a lysophosphatidylethanolamine metabolite supplied as a high-purity reference standard for research applications, including biomarker studies and analytical method development. It is used in metabolomics and toxicity-assessment workflows to support identification and quantitation of lysophospholipids.
  • High reported purity suitable for analytical use.
  • Supplied in small quantities appropriate for method development and standards.
  • Molecular weight and formula provided for accurate mass calculations.
  • Serves as a biomarker reference in toxicity and metabolomics studies.
  • Solid form with defined storage recommendations to maintain stability.
Catalog No. 50-002-87220