Learn More
Medchemexpress LLC LysoPE(18:2/0:0) | 85046-18-0 | C23H44NO7P | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1549181MG
Encompass_Preferred
LysoPE (18:2/0:0) is a lysophosphatidylethanolamine involved in phospholipid metabolism, with significantly altered serum levels observed in mice exposed to DEHP and Aroclor 1254. It is suitable for research concerning endocrine-disrupting compound-induced metabolic disorders and lipid metabolism disturbance, and can serve as a potential biomarker for combined toxicity studies.
  • Involved in phospholipid metabolism
  • Potential biomarker for combined toxicity of DEHP and Aroclor 1254
  • Useful for research into metabolic disorders and lipid metabolism disturbance
  • Appears as a white to off-white solid with 99.59% purity
Catalog No. 50-003-76200