Learn More
Enzo Life Sciences Lysophosphatidic acid, (5mg), CAS Number: 325465-93-8
SDP
Supplier:  Enzo Life Sciences BMLLP1000005
Encompass_Preferred
LPA receptor ligand. Lysophosphatidic acid (LPA) is a multifunctional intercellular phospholipid messenger. LPA stimulates the growth of a variety of cells including fibroblasts, vascular smooth muscle cells, endothelial cells and keratinocytes. It acts as a proliferative and anti-apoptotic factor. Ligand for LPA1 (EDG-2), LPA2 (EDG-4) and LPA3 (EDG-7) receptors. It also inhibits differentiation of neural stem cells into neurons. Alternative Name: 1-Oleoyl-lysophosphatidic acid . sodium salt. Formula: C21H40O7P . Na. MW: 458.5. CAS Number: 325465-93-8. Purity: ≥98% (TLC). Appearance: Waxy solid. Solubility: Soluble in water (5 mg/mL with sonication) or DMSO (0.5 mg/mL warm). Use/Stability: Store as supplied at -20°C for up to 1 year. Store solutions at -20°C for up to 1 month. Long Term Storage: -20°C
Catalog No. NC1657890