Learn More
Medchemexpress LLC Lysophosphatidylcholine 18:2 | 22252-07-9 | 99.1% | 519.65 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN941010MG
Encompass_Preferred
Lysophosphatidylcholine 18:2 is a lysophospholipid and a potential biomarker identified from insulin resistance (IR) polycystic ovary syndrome (PCOS). Low plasma levels of this compound predict impaired glucose tolerance, insulin resistance, type 2 diabetes, coronary artery disease, and memory impairment. This product is for research use only and not sold to patients.
  • Lysophospholipid and endogenous metabolite
  • Potential biomarker for insulin resistance (IR) polycystic ovary syndrome (PCOS)
  • Associated with impaired glucose tolerance, insulin resistance, type 2 diabetes
  • Implies inflammation, obesity, and memory impairment
  • Contributes to the separation of IR PCOS and control groups
  • For research use only
Catalog No. 50-004-26201