Learn More
Medchemexpress LLC Lysophosphatidylcholine 18:2 | 22252-07-9 | 99.1% | 519.65 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN94105MG
Encompass_Preferred
Lysophosphatidylcholine 18:2 (1-Linoleoyl-2-Hydroxy-sn-glycero-3-PC) is a lysophospholipid identified as a potential biomarker for insulin resistance (IR) polycystic ovary syndrome (PCOS). Low plasma levels of this compound are associated with impaired glucose tolerance, insulin resistance, type 2 diabetes, coronary artery disease, and memory impairment. It is classified as a human endogenous metabolite and is relevant to disease markers, infection, and endocrine diseases.
  • Contributes to distinguishing between IR PCOS and control groups
  • Associated with inflammation, insulin resistance, obesity, and type 2 diabetes
  • A major class of glycerophospholipids in human plasma
Catalog No. 50-004-25537