Learn More
Medchemexpress LLC Lysophosphatidylethanolamine | 85046-18-0 | C23H44NO7P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1549185MG
Encompass_Preferred
LysoPE (18:2/0:0) is a lysophosphatidylethanolamine involved in phospholipid metabolism. It shows significantly altered serum levels in mice exposed to a combination of DEHP and Aroclor 1254, which is linked to disturbed phospholipid metabolism. This compound is useful for research on endocrine-disrupting compound-induced metabolic disorders and lipid metabolism disturbance, and has been identified as a potential biomarker for the combined toxicity of DEHP and Aroclor 1254.
  • Involved in phospholipid metabolism
  • Serum levels altered by exposure to DEHP and Aroclor 1254
  • Useful for research on endocrine-disrupting compound-induced metabolic disorders
  • Identified as a potential biomarker for combined toxicity of DEHP and Aroclor 1254
Catalog No. 50-003-76920