Learn More
Sigma Aldrich Fine Chemicals Biosciences Lysosome Isolation Kit suf
Supplier:  Sigma Aldrich Fine Chemicals Biosciences LYSISO11KT
Encompass
Lysosomes are organelles ubiquitous in most prokaryotic and eukaryotic cells. They contain many acid hydrolases that take part in protein degradation in the cell. Lysosomes also contain lipases nucleases and polysaccharidases and deficiencies in some of these enzymes lead to specific lysosomal storage diseases such as Tay Sachs Gaucher Hunter disease and others. They also contribute to maintaining cellular homeostasis. Malfunctions in this organelle directly impact cell behavior and fate. Lysosomes may also be involved in other cellular processes such as Albinism and aging.
Catalog No. 50-290-7927