Learn More
Biotium LysoView 640, 50 μL, far-red fluorescent dye, no-wash lysosome staining, 1000X in DMSO, 640 nm ex, 671 nm em
Supplier:  Biotium 70085
Encompass_Preferred
LysoView™ 640 (50 uL) is a far-red fluorescent dye designed for efficient staining and imaging of lysosomes in live cells. This reagent is ideal for real-time live-cell imaging, accumulating within low pH acidic organelles to enable clear visualization of lysosome localization and morphology. Its excellent photostability and non-toxic properties allow for prolonged use without harm to cells.
  • Efficient accumulation in acidic organelles
  • Real-time live-cell imaging
  • Excellent photostability
  • Non-toxic for live-cell use
  • Compatible with adherent and suspension cells
  • No-wash staining protocol
  • Store at -20°C, protect from light
  • At least 12 months shelf life
  • Not well retained after formaldehyde fixation
Catalog No. NC2175051