Learn More
Medchemexpress LLC Lysozyme chloride | 9066-59-5 | 1 G

Supplier: Medchemexpress LLC HYB2237A1G
Lysozyme chloride is a bactericidal enzyme that lyses gram-positive bacteria. It can be used for the research of HIV infection and pulmonary emphysema. This natural antimicrobial hydrolyzes the β(1-4) glycosidic linkage in the peptidoglycan layer of the bacterial cell wall, causing cell lysis. Its bactericidal effect is primarily limited to gram-positive bacteria.
- Appearance: Solid
- Color: White to off-white
- Solubility: ≥ 4 mg/mL in H2O
- Targets bacteria and HIV
- Anti-infection pathway
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.