Learn More
Medchemexpress LLC Lysozyme chloride | 9066-59-5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB2237A100MG
Encompass_Preferred
Lysozyme chloride is a bactericidal enzyme that lyses gram-positive bacteria. It can also be used for research on HIV infection and pulmonary emphysema. This natural antimicrobial hydrolyzes the β(1-4) glycosidic linkage in the peptidoglycan layer of bacterial cell walls, leading to cell lysis. Its bactericidal effect is primarily against gram-positive bacteria, including pathogens like *Listeria monocytogenes* and certain *Clostridium* species, as well as some spoilage organisms.
  • Lyses gram-positive bacteria.
  • Useful for research on HIV infection.
  • Useful for research on pulmonary emphysema.
  • Hydrolyzes β(1-4) glycosidic linkage in bacterial cell walls.
  • Effective against pathogens like *Listeria monocytogenes* and certain *Clostridium* species.
  • Can impair hyaluronan's ability to prevent elastase injury to elastic fibers.
Catalog No. 50-004-09725