Learn More
Cusabio Technology LLC Fish lysozyme (renal amyloidosis) (LZM) ELISA kit
Supplier:  Cusabio Technology LLC CSBE17296FH
Encompass_Preferred
This Fish LZM ELISA Kit was designed for the quantitative measurement of Fish LZM protein in serum, plasma. It is a Competitive ELISA kit, its detection range is 3.12 ng/mL-200 ng/mL and the sensitivity is 1.56 ng/mL. One kit contains 96 wells.
Catalog No. NC2952970