Learn More
AG Scientific Inc Lyticase 100T, 25 MG
Supplier:  AG Scientific Inc L510025MG
Encompass_Preferred
Lyticase, produced by a submerged culture of Arthrobacter luteus, is an enzyme preparation which effectively lyses cell walls of viable yeast cells. An essential enzyme responsible for lysis of viable yeast cells in this preparation is b-1, 3-glucan laminaripentaohydrolase.It hydrolyzes linear glucose polymers with b-1,3-linkages and releases laminaripentaose specifically as the main and minimum product unit (4,5,10,11). This lytic activity releases spheroplasts and protoplasts in the preparation of yeast DNA prior to restriction enzyme digestion and Southern Blot analysis. The extent of lysis of yeast cells by lyticase varies with yeast strain, growth stage of yeast and cultural condition.CAS Number: 37340-57-1Storage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6954