Learn More
Medchemexpress LLC m-3M3FBS | 200933-14-8 | 99.99% | 343.36 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1961910MM/1ML
Encompass_Preferred
m-3M3FBS is a potent phospholipase C (PLC) activator that stimulates superoxide generation in human neutrophils. It upregulates intracellular calcium concentration and stimulates inositol phosphate generation in various cell lines. Additionally, m-3M3FBS induces apoptosis in monocytic leukemia cells.
  • Potent phospholipase C (PLC) activator.
  • Stimulates superoxide generation in human neutrophils.
  • Upregulates intracellular calcium concentration.
  • Stimulates inositol phosphate generation in various cell lines.
  • Induces monocytic leukemia cell apoptosis.
Catalog No. 50-004-01263