Learn More
Medchemexpress LLC m-3M3FBS | 200933-14-8 | 99.99% | 343.36 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1961950MG
Encompass_Preferred
m-3M3FBS is a potent phospholipase C (PLC) activator. It stimulates superoxide generation in human neutrophils, upregulates intracellular calcium concentration, and stimulates inositol phosphate generation in various cell lines. It also induces monocytic leukemia cell apoptosis.
  • Stimulates superoxide generation in human neutrophils
  • Upregulates intracellular calcium concentration
  • Stimulates inositol phosphate generation in various cell lines
  • Induces monocytic leukemia cell apoptosis
Catalog No. 50-004-00472