Learn More
Medchemexpress LLC M-3m3fbs | 200933-14-8 | MFCD00095824 | 100.0% | 343.36 | C16H16F3NO2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1961910MG
Encompass_Preferred
m-3M3FBS is a small-molecule research reagent that functions as a potent phospholipase C (PLC) activator for in vitro and in vivo studies. It is supplied as a white to off-white solid with high reported purity, documented solubility in DMSO, and recommended storage conditions for laboratory use.
  • Potent phospholipase C activator suitable for functional assays.
  • High reported purity for reproducible experimental results.
  • Soluble in DMSO at high concentration; in vivo formulation protocols provided.
  • Stable as a powder at low temperature and in solution when stored frozen.
  • Available in small pack sizes for screening and dose-finding studies.
Catalog No. 50-002-98372