Learn More
Medchemexpress LLC M-BBIG dihydrochloride | 181147-28-4 | 98.2% | 319.19 | 50 MG

Supplier: Medchemexpress LLC HY16465250MG
m-BBIG dihydrochloride is a member of the Bis-iminoguanidine (BIG) family, utilized in synthetic chemistry. This high-purity compound is provided as a white to off-white solid.
- Member of the Bis-iminoguanidine (BIG) family
- Suitable for synthetic chemistry applications
- Purity of 98.21%
- Appears as a white to off-white solid
- Chemical formula: C10H16Cl2N8
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.