Learn More
Zymo Research Corporation M-Binding Buffer, 250 ml
SDP
Supplier:  Zymo Research Corporation D50403
Encompass_Preferred
Used for purification of bisulfite converted DNA on a Zymo-spin column in the EZ DNA Methylation, EZ DNA Methylation - Gold, EZ DNA Methylation - Direct kits, and EZ DNA Methylation - Lightning kits.
Catalog No. 50-203-7123