Learn More
Bellco Glass, Inc. MicroCarrier Spinner Only, 500mL,70mm Nk W/Dimple Bottom - No Ctr Cap
SDP
Supplier:  Bellco Glass, Inc. 196510500
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 500ml Sidearm cap: 32mm (included)
Catalog No. 50-194-4979