Learn More
Bellco Glass, Inc. MicroCarrier Spinner Flask Only, 8L No Ctr Cap
SDP
Supplier:  Bellco Glass, Inc. 196518000
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 8l Sidearm cap: 45mm (included) •100mm ctr neck•(2) 45mm angled sidearms (30°)•dimensions: 238mm od x 420mm overall height•graduations: 1500ml to 8000ml - every 500ml•supply with two (2) 45mm pp blue s/caps & pouring rings•supply with no center cap (for center solid cap; A523-146)•material: type 1 class a borosilicate glass•weight of flask (empty): 8lbs (approx)
Catalog No. 50-194-4988