Learn More
Medchemexpress LLC M-carboxycinnamic acid bishydroxamide | 174664-65-4 | MFCD03453553 | 98.0% | 222.20 g·mol^-1 | C10H10N2O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW0140045MG
Encompass_Preferred
m-Carboxycinnamic acid bishydroxamide (CBHA) is a small-molecule histone deacetylase (HDAC) inhibitor supplied for research use. It is used in biochemical and cellular assays to probe chromatin regulation, gene expression, and apoptosis pathways, with reported activity against HDAC1 and HDAC3.
  • potent HDAC1 and HDAC3 inhibition (reported ID50 ≈ 10 nM and 70 nM)
  • high purity suitable for biochemical assays (98.0%)
  • available in small sample sizes for screening and method development
  • stable as a powder when stored at low temperature
  • compatible with in vitro cellular and enzymatic studies
Catalog No. 50-003-03815