Learn More
Abcam m-Cresol purple, pH indicator dye, 25G
Supplier:  Abcam AB14631825G
Encompass_Preferred
MW 382.43 g/mol. pH indicator dye used primarily in spectrophotometric pH measurements. Acidic and hydrophobic. The useful pH ranges are 1.2 - 2.8 (red to yellow) and 7.4 - 9.0 (yellow to violet).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4728