Learn More
Medchemexpress LLC M-CSF Human CHO 2ug
SDP
Supplier:  Medchemexpress LLC HYP7050A2UG
Encompass_Preferred
Recombinant human macrophage colony-stimulating factor (M-CSF) produced in CHO cells and supplied as a lyophilized powder for use in cell culture and biochemical assays to stimulate macrophage proliferation, differentiation, and CSF-1 receptor signaling studies.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Supplied as lyophilized powder for convenient storage and handling.
  • Bioactive with ED50 <5 ng/mL in cell proliferation assays using M-NFS-60 cells.
  • Glycosylated protein showing expected molecular weight shift, suitable for functional studies.
  • Stable when stored at -20°C; aliquoting recommended for long-term storage.
  • Recommended reconstitution at ≥100 μg/mL and use of carrier protein for extended stability.
Catalog No. 50-001-84812