Learn More
Medchemexpress LLC M-CSF Rat 2ug
SDP
Supplier:  Medchemexpress LLC HYP73862UG
Encompass_Preferred
M-CSF Protein Rat is a pro-inflammatory cytokine binds to its receptor CSF1R and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts which are important in the destruction of bone and cartilage and in the periarticular osteoporotic changes
Catalog No. 50-001-79342