Learn More
Medchemexpress LLC M-CSF Rat CHO 100ug
SDP
Supplier:  Medchemexpress LLC HYP7247100UG
Encompass_Preferred
M-CSF Protein Rat (CHO) is a pro-inflammatory cytokine which binds to its receptor CSF1R and is involved in the development and proliferation of cells of the monocyte/macrophage lineage and participates in the induction of osteoclasts
Catalog No. 50-001-82578