Learn More
Apexbio Technology LLC m-Hydroxybenzoic acid 99-06-9 5g
SDP
Supplier:  Apexbio Technology LLC B79025000
Encompass_Preferred
m-Hydroxybenzoic acid (CAS nan) is a small molecule intermediate utilized in the synthesis of pharmacologically active compounds It serves as a precursor for generating derivatives including kinase inhibitors targeting Tie-2 signaling and amyloidogenesis modulators In biomedical research m-hydroxybenzoic acid is primarily employed as a starting material to design small molecule analogs such as oxadiazole-based compounds structurally associated with resveratrol for antioxidant and anti-inflammatory research applications Based on these pharmacological properties m-hydroxybenzoic acid holds research potential in the development of kinase inhibitors amyloid-related modulators and antioxidant or anti-inflammatory agents
Catalog No. 50-000-91158