Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-methyl red | 20691-84-3 | MFCD00059641 | 98.0% | 269.30 g/mol | C15H15N3O2 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW1301285G

Encompass_Preferred

m-Methyl red is an azo dye and pH-sensitive acid-base indicator supplied as a solid for laboratory and analytical applications, including titration endpoint detection and staining in research settings.

  • Indicator: pH transition around 4.4-6.2 (pKa ≈5.1)
  • Useful for visual titration endpoint detection
  • Chemical formula C15H15N3O2 and molecular weight 269.30 g/mol
  • Provided as a solid suitable for staining and analytical uses
  • CAS number 20691-84-3 for precise chemical identification

Catalog No. 50-002-82622


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.