Learn More
Medchemexpress LLC M-MLV Reverse transcriptase | 9068-38-6 | 1 KU
SDP
Supplier:  Medchemexpress LLC HY1311611KU
Encompass_Preferred
M-MLV Reverse transcriptase (M-MLV RT) is an enzyme derived from the Moloney murine leukemia virus. It is primarily used in scientific research and drug registration applications.
  • Activity: 200 U/μL.
  • Unit defined as the incorporation of 1 nmol of dTTP into an acid-insoluble fraction.
  • cDNA products can be stored at -20°C or -80°C, or used immediately for PCR (qPCR) reactions.
Catalog No. 50-003-38809