Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-MLV Reverse transcriptase | 9068-38-6 | 5 KU
SDP

Supplier:  Medchemexpress LLC HY1311615KU

Encompass_Preferred

M-MLV Reverse transcriptase is a colorless to light yellow liquid with an activity of 200 U/μL. One unit is defined as the amount of enzyme required to incorporate 1 nmol of dTTP into an acid-insoluble fraction using poly(rA)•oligo(dT) as the template-primer under 37 °C for 10 min.

  • Source: E.coli
  • Store at -20°C for 2 years
  • For research use only

Catalog No. 50-003-41144


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.