Learn More
Medchemexpress LLC m-PEG-DMG | 1019000-64-6 | 98.0% | 2000 (average) | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW44082150MG
Encompass_Preferred
m-PEG-DMG (MW 2000) is a PEGylated lipid supplied as a powder with an average PEG molecular weight of 2000. It is intended for incorporation into liposomes and lipid nanoparticles for formulation and drug delivery research, providing a PEG surface modification to improve colloidal stability and in vivo circulation.
  • PEGylated lipid suitable for liposome and lipid nanoparticle formulation.
  • Average molecular weight about 2000 for consistent PEG chain length.
  • Enhances particle stability and circulation time in biological systems.
  • Supplied as a powder compatible with solvent-based formulation workflows.
  • Stable under recommended frozen storage for extended shelf life.
Catalog No. 50-002-81683