Learn More
Medchemexpress LLC m-PEG-DMG | 1019000-64-6 | 98.0% | 2000 Da (average) | (C2H4O)nC36H73NO5 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW440821100MG
Encompass_Preferred
m-PEG-DMG (MW 2000) is a methoxy-terminated polyethylene glycol conjugated to dimyristoylglycerol. Supplied as a solid with an average molecular weight of about 2000 Da, it is formulated for preparation of PEGylated liposomes and lipid nanoparticles used in drug delivery research and related in vitro studies.
  • High purity (98.0%).
  • Average molecular weight ~2000 Da.
  • White to off-white solid form.
  • Suitable for liposome and lipid nanoparticle formulation.
  • Provides steric stabilization and reduced protein opsonization.
  • Available in small laboratory pack sizes (e.g., 5-100 mg).
  • Documentation available: data sheet, COA, and SDS.
Catalog No. 50-002-81682