Learn More
Medchemexpress LLC m-PEG-DMG | 5,000 Da (average) | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16698310MG
Encompass_Preferred
m-PEG-DMG (MW 5000) is a methoxy-terminated polyethylene glycol (PEG) conjugated to a dimyristoyl glycerol (DMG) lipid tail used as a PEG-lipid component in lipid nanoparticle and drug delivery formulations. It provides steric stabilization and can improve the delivery efficiency and tissue specificity of poorly soluble drugs.
  • Used as a PEG-lipid in lipid nanoparticles and drug delivery research.
  • Average molecular weight about 5,000 Da.
  • Typically supplied as a powder with recommended storage: powder -20°C; in solvent -80°C for 6 months or -20°C for 1 month.
  • Solubility in DMSO approximately 50 mg/mL (ultrasonic assistance may be required).
  • Provides steric stabilization to nanoparticle surfaces and can reduce aggregation and opsonization.
Catalog No. 50-002-89188