Learn More
Medchemexpress LLC m-PEG-DSPE sodium | 95.0% | 3000 Da (average) | (C2H4O)nC44H88NO9P | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW440924500MG
Encompass_Preferred
m-PEG-DSPE sodium (MW 3000) is a PEGylated phospholipid sodium salt supplied as a solid. It is commonly used to produce long-circulating liposomes and lipid nanoparticles to improve colloidal stability, steric protection, and tissue distribution of poorly soluble drugs. Manufacturer documentation includes a datasheet, certificate of analysis, and safety data sheet.
  • PEGylated phospholipid with average molecular weight ~3000 Da.
  • Improves nanoparticle colloidal stability and steric hindrance.
  • Prolongs circulation time in vivo.
  • Compatible with liposome and lipid nanoparticle formulations.
  • Available as a solid in 500 MG pack sizes.
  • Typical purity approximately 95.0%.
Catalog No. 50-002-81685