Learn More
Medchemexpress LLC m-PEG-DSPE sodium (MW 3000) | 95.0% | 3,000 Da | 50 MG

Supplier: Medchemexpress LLC HYW44092450MG
m-PEG-DSPE sodium (MW 3000) is a methoxy-polyethylene glycol-conjugated distearoylphosphatidylethanolamine sodium salt intended for research use in drug delivery and nanoparticle or liposome surface modification. It is a PEGylated lipid (PEG average MW 3000) provided as a purified polymeric lipid material suitable for formulation and surface-stabilization applications.
- Provides PEGylation for improved circulation and reduced opsonization.
- Average PEG molecular weight 3,000 Da for steric stabilization.
- Sodium salt form for aqueous compatibility.
- Purity approximately 95.0%.
- Suitable for nanoparticle, liposome, and formulation research.
- Available in small laboratory quantities such as 50 mg and larger sizes by request.
- Store sealed away from moisture; in solvent: -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.