Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC m-PEG-NH2 hydrochloride | 99.1% | 2000 Da (average) | Polydisperse PEG (repeating unit C2H4O) hydrochloride (no single chemical formula) | 25 MG
SDP

Supplier:  Medchemexpress LLC HYW598230A25MG

Encompass_Preferred

Amine-terminated methoxy polyethylene glycol hydrochloride with an average molecular weight of 2000 Da. This polydisperse PEG reagent is used as a building block and barrier-permeation modifier for preparing hybrid gels and for size-exclusion applications in biochemical assays.

  • Amine-terminated methoxy polyethylene glycol hydrochloride with average molecular weight 2000 Da.
  • Polydisperse polymeric reagent suitable for gel and biochemical assay applications.
  • Helps AdSEC gels separate from complex biological mixtures such as blood, urine, sweat, and tears.
  • Reported purity 99.13%.
  • Soluble in DMSO at 100 mg/mL (ultrasonic recommended).
  • Store at -20°C and protect from light; in solvent store at -80°C for up to 6 months or -20°C for 1 month.

Catalog No. 50-002-84002


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.