Learn More
Medchemexpress LLC m-PEG-NHS ester | 92451-01-9 | MFCD06412227 | 90.0% | 350 g/mol | (C2H4O)nC7H9NO5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY155909C10MG
Encompass_Preferred
m-PEG-NHS ester (MW 350) is a methoxy-terminated polyethylene glycol activated as an N-hydroxysuccinimide (NHS) ester for covalent PEGylation of primary amine-containing molecules. It is intended to improve solubility and stability, reduce immunogenicity, and enable bioconjugation and surface functionalization in research applications.
  • Activated NHS ester for efficient coupling to primary amines.
  • Methoxy termination prevents crosslinking and limits multi-attachment.
  • Improves solubility and stability of biomolecules.
  • Reduces immunogenicity and can extend circulation half-life.
  • Suitable for protein, peptide, and small-molecule PEGylation.
  • Available in small mg-scale quantities for research use.
Catalog No. 50-002-88124