Learn More
Medchemexpress LLC M-PEG-NHS ester (MW 20000) | 92451-01-9 | 98.0% | (C2H4O)nC7H9NO5 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140700
Encompass_Preferred
M-PEG-NHS ester (MW 20000) is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds that consist of two different ligands connected by a linker; one ligand targets an E3 ubiquitin ligase, and the other targets the protein of interest. This mechanism allows PROTACs to utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52678