Learn More
Medchemexpress LLC m-PEG-OH (MW5000) | 9004-74-4 | 98.0% | 500 G
SDP
Supplier:  Medchemexpress LLC HY140697500G
Encompass_Preferred
m-PEG-OH (MW5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules connect two ligands, one for an E3 ubiquitin ligase and another for a target protein, enabling the selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Exploits the intracellular ubiquitin-proteasome system
Catalog No. 50-003-56630