Learn More
Medchemexpress LLC m-PEG-thiol | 134874-49-0 | MFCD00803682 | 90.0% | 750 (average) | (C2H4O)nC3H8OS | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW591476B250MG
Encompass_Preferred
m-PEG-thiol (average MW 750) is a methoxy-terminated polyethylene glycol bearing a terminal thiol group, supplied as a solid reagent for bioconjugation and nanoparticle surface functionalization. It is commonly used to thiolate DNA and to anchor polymers or biomolecules onto gold and other metal surfaces via thiol-gold chemistry, supporting applications in nanostructure assembly, biosensing, and drug delivery.
  • Provides terminal thiol for covalent attachment to metal surfaces and thiol-reactive chemistries.
  • Average molecular weight 750 for a short PEG spacer length.
  • Supplied as a solid suitable for solution-phase conjugation.
  • Reported purity approximately 90%.
  • Compatible with DNA thiolation and nanoparticle surface modification protocols.
Catalog No. 50-002-84500