Learn More
Medchemexpress LLC M-PEG-thiol (MW 2000) | 134874-49-0 | 99.8% | COCCOCCS.[MW 2000].[n] | 1 G
SDP
Supplier:  Medchemexpress LLC HY1407091G
Encompass_Preferred
m-PEG-thiol (MW 2000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. This product is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-56634