Learn More
Medchemexpress LLC M-PEG-triethoxysilane (MW 5000) | 98.0% | COCCOCCNC(NCCC[Si](OCC)(OCC)OCC)=O.[n].[MW 5000] | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140704100MG
Encompass_Preferred
M-PEG-triethoxysilane (MW 5000) is a PEG-based PROTAC linker intended for research use only. It can be utilized in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • PROTACs exploit the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53516