Learn More
Medchemexpress LLC m-PEG1-NHS ester 5g | 1027371-75-0 | 201.18 g/mol | C8H11NO5 | 5 G
SDP
Supplier:  Medchemexpress LLC HY1145125G
Encompass_Preferred
m-PEG1-NHS ester is a methoxy-terminated PEG1 N-hydroxysuccinimide ester that provides an activated NHS ester for efficient coupling to primary amines. It is used as a short PEG linker in bioconjugation and PROTAC synthesis to introduce a flexible spacer and enable amide bond formation under mild conditions.
  • Provides a methoxy-terminated PEG1 spacer for minimal linker length.
  • Contains an active NHS ester for rapid amide coupling to primary amines.
  • Suitable for PROTAC linker assembly and general bioconjugation.
  • Available in multiple pack sizes for flexible scaling.
Catalog No. 50-002-30058