Learn More
Medchemexpress LLC m-PEG11-amine | 854601-60-8 | 515.64 g/mol | C23H49NO11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW040222100MG
Encompass_Preferred
m-PEG11-amine is a PEG-based linker bearing a terminal amine used as a cleavable linker in antibody-drug conjugates (ADCs) and as a PROTAC linker or PEG spacer in conjugation chemistry. It provides a water-soluble spacer that facilitates conjugation to activated esters, carboxylic acids, and other reactive groups, improving solubility and flexibility of the resulting bioconjugates.
  • PEG-based linker with a terminal primary amine for conjugation
  • Used as a cleavable linker in antibody-drug conjugates (ADCs)
  • Suitable as a PROTAC linker or PEG spacer in conjugation chemistry
  • Water-soluble spacer improves solubility and flexibility of conjugates
  • Available in multiple sizes for research-scale synthesis
Catalog No. 50-002-03088