Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC M-PEG12-amine | 1977493-48-3 | MFCD11041151 | ≥98.0% | 559.69 g/mol | C25H53NO12 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY14022725MG

Encompass_Preferred

m-PEG12-amine is a monodisperse polyethylene glycol (PEG) linker bearing a terminal primary amine, used for bioconjugation, PROTAC synthesis, and as a non-cleavable 12-unit PEG spacer for antibody-drug conjugates.

  • Monodisperse 12-unit PEG spacer
  • Terminal primary amine for conjugation
  • Molecular weight 559.69 g/mol
  • Molecular formula C25H53NO12
  • Purity ≥98.0% (as stated on COA)

Catalog No. 50-002-02889


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.